Stampa…

Anti-MAT1/2A Rabbit Polyclonal Antibody (DyLight® 405)

Fornitore: Novus Biologicals
NB110-94162V
NOVUNB110-94162VEA 722 EUR
NOVUNB110-94162V
Anti-MAT1/2A Rabbit Polyclonal Antibody (DyLight® 405)
Anticorpi
Type: Primary
Antigen: MAT1/2A
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR