Stampa…

Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody (DyLight® 550)

Fornitore: Novus Biologicals
NB110-93479R
NOVUNB110-93479REA 718 EUR
NOVUNB110-93479R
Anti-Pyruvate Dehydrogenase E1-alpha subunit [p Ser293] Rabbit Polyclonal Antibody (DyLight® 550)
Anticorpi
Type: Primary
Antigen: Pyruvate Dehydrogenase E1-alpha subunit [p Ser293]
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR