Stampa…

Anti-Perilipin Rabbit Polyclonal Antibody

Fornitore: Novus Biologicals
NB110-40760 NB110-40760SS
NOVUNB110-40760EA 606 EUR
NOVUNB110-40760 NOVUNB110-40760SS
Anti-Perilipin Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: Perilipin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR