Stampa…
Covers for equipment and glassware, BioClean™
Fornitore: Nitritex
BPC-122 BPC-091 BPC-229 BPC-183 BPC-152 BPC-061 BPC-018 BPC-051 BPC-041 BPC-030 BPC-023 BPC-013 BPC-010 BPC-008 BPC-076
NITRBPC-122EA 0 EUR
NITRBPC-122 NITRBPC-091 NITRBPC-229 NITRBPC-183 NITRBPC-152 NITRBPC-061 NITRBPC-018 NITRBPC-051 NITRBPC-041 NITRBPC-030 NITRBPC-023 NITRBPC-013 NITRBPC-010 NITRBPC-008 NITRBPC-076
Covers for equipment and glassware, BioClean™
Protezione delle superfici Dispositivi di protezione per strumentazione
These BioClean™ covers provide protection from environmental impurities during storage, use and transportation. They also minimise the need for autoclave tape during wrapping and save time when prepping glassware or tubing for autoclaving.
  • Rubber elastic provides secure seal
  • Lint-free, moisture and puncture resistant
  • Fast drying
  • Easy to apply and remove
  • Autoclavable at 121 ºC
Certificazioni: Manufactured in a facility holding ISO 9001: 2008 certification.
Confezione: 50 pieces per sealed inner PE bag; 10 inner bags per sealed outer PE bag; OneMore Product Information