Stampa…
Absorbent mat pads for viscous liquids, PIG®
Fornitore: NEW PIG
MAT209
NEWPMAT209EA 338 EUR
NEWPMAT209
Absorbent mat pads for viscous liquids, PIG®
Materiali per il controllo di fuoriuscite di liquidi
White polyester fibre absorbent mat pads for viscous liquids for picking up paint, glue, resins, shampoo, liquid chocolate, syrup, varnish.
Up to 3,63 litres of viscous fluids per pad.