Stampa…
Flaconi diagnostici, collo stretto, rotondi, senza tappo a vite Nalgene®
Fornitore: Thermo Fisher Scientific
362006-9050 362006-9025 362006-9125
NALG362006-9050EA 1271 EUR
NALG362006-9050 NALG362006-9025 NALG362006-9125
Flaconi diagnostici, collo stretto, rotondi, senza tappo a vite Nalgene®
Bottiglie Bottiglie in plastica
Bottles without closure use in sample storage and the shipping of small volume, valuable reagents.
  • Nonsterile
  • Natural translucent PPCO provides excellent chemical resistance
  • Durable and feature uniform walls resistant to splitting or puncturing
  • Leakproof when assembled with closure
Certificazioni: Compliance with ISO 13485:2003 and 21CFR177.1520, USP Class VI.
Informazioni per l'ordine: Coloured closures are ordered seperately.