Stampa…
Bottiglie a collo largo, HDPE, rotonde, con tappo a vite, IP2
Fornitore: Thermo Fisher Scientific
312199-0004 312199-0008 312199-0016 312199-0032
NALG312199-0004EA 1890 EUR
NALG312199-0004 NALG312199-0008 NALG312199-0016 NALG312199-0032
Bottiglie a collo largo, HDPE, rotonde, con tappo a vite, IP2
Bottiglie Bottiglie in plastica
Wide-mouth HDPE IP 2 bottles, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof when used with Nalgene closures
  • Round shape
Safe, durable, economical bottles for the transport and packaging of hazardous goods. Recommended for customers who wish to put together and certify their own packaging system. Extra-wide mouth for simple filling and emptying of liquids and dry substances. The sturdy walls withstand cracking and damage from impact caused by shocks during transport. The bottom of the bottleMore Product Information