Stampa…
Bottiglie, a bocca stretta, fluorinate, con tappo a vite, Nalgene®
Fornitore: Thermo Fisher Scientific
312097-0004 312097-0008 312097-0016 312097-0032
NALG312097-0004EA 0 EUR
NALG312097-0004 NALG312097-0008 NALG312097-0016 NALG312097-0032
Bottiglie, a bocca stretta, fluorinate, con tappo a vite, Nalgene®
Bottiglie Bottiglie in plastica
HDPE, fluorinated with PP, fluorinated screw cap.
  • Fluorinated bottles and carboys can be used for most acids, alkalis and aggressive organic solvents
  • Round shape, non sterile
  • Heavy-duty walls are safe, durable and resistant to splitting and punctures
  • Fluorocarbon surfaces inside and out
  • Leakproof
Fluorinating these HDPE bottles and carboys provides a cost effective alternative to costly fluoropolymer containers.
Manufactured from Nalgene™ for resistance to permeation, paneling, odor emission, flavor and fragrance loss. Provide superior recepticalsMore Product Information
Visualizza i codici
Specifiche
Environmentally Preferable Produzione a basso impatto
Certificazione ambientale NAR Certificate of Retirement