Stampa…
Borracce, collo largo, ambra, Nalgene®
Fornitore: Thermo Fisher Scientific
2256-7020
NALG2256-7020EA 136 EUR
NALG2256-7020
Borracce, collo largo, ambra, Nalgene®
Bottiglie Bottiglioni
Made of HDPE, amber.
  • Good chemical resistance
  • Good temperature resistance
Protect photosensitive chemicals, reagents, buffers and standards when mixing and storing. Amber HDPE carboys with amber polypropylene (PP) closure meet the U.S. Pharmacopoeia Light Transmission Test (USP latest edition) requirements for storage of light sensitive materials.