Stampa…
Safety Storage Cabinet, Type 90, ION-CORE-90 CHARGE model IO90.195.060.CC.WDC
Fornitore: asecos
41373 41302047 41366 41369
MSGA41373EA 2590 EUR
MSGA41373 135-2885 MSGA41366 MSGA41369
Safety Storage Cabinet, Type 90, ION-CORE-90 CHARGE model IO90.195.060.CC.WDC
Arredi Armadietti per la conservazione
Safe active and passive storage of lithium-ion batteries.
  • Robust construction and longevity
  • Integrated potential-free alarm contact
  • No unauthorised use
  • Integrated transport base
  • Easy alignment
  • Smoke detector
  • Pressure relief
  • All-around protection
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface. Integrated potential-free alarm contact for strongly recommended alarm forwarding. Optionally available with module for remote signalling.
No unauthorised use: door lockable withMore Product Information