Stampa…
Safety Storage Cabinet, Type 90, ION-CORE-90 STORE model IO90.195.060.CS.WDC
Fornitore: asecos
41362 41419047 41363 41364 41365
MSGA41362EA 418 EUR
MSGA41362 135-2883 MSGA41363 MSGA41364 MSGA41365
Safety Storage Cabinet, Type 90, ION-CORE-90 STORE model IO90.195.060.CS.WDC
Arredi Armadietti per la conservazione
Safe passive storage of lithium-ion batteries.
  • Robust construction and longevity
  • Integrated potential-free alarm contact
  • No unauthorised use
  • Integrated transport base
  • Easy alignment
  • Smoke detector
  • Pressure relief
  • All-around protection
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface. Integrated potential-free alarm contact for strongly recommended alarm forwarding. Optionally available with module for remote signalling.
No unauthorised use: door lockable with cylinder locking (More Product Information