Stampa…
Blood glucose lancets, VITREX® Soft
Fornitore: Vitrex Medical A/S
351213
MODU351213EA 15 EUR
MODU351213
Blood glucose lancets, VITREX® Soft
Lancette pungidito
Needle material is stainless steel grade 304/1 Cr18Ni9 and moulded body/cap material is LDPE (Low Density Polyethylene).
  • Good blood flow - Triple ground needle point
  • Sterilised by gamma irradiation at minimum 25kGy
  • Convenient boxes with dispensing feature
VITREX® Soft is designed for use with most popular types of finger lancing device.