Stampa…
Scotch-Brite™ 2010 High Performance Microfibre Wipes
Fornitore: 3M
2010HLWS 2010HLGN2 2010HLRT2 SB2010YE SB2010HPW
MMMA2010HLWSEA 65.1 EUR
MMMA2010HLWS MMMA2010HLGN2 MMMA2010HLRT2 MMMASB2010YE MMMASB2010HPW
Scotch-Brite™ 2010 High Performance Microfibre Wipes
Panni Panni per pulizia riutilizzabili Panni per operazioni delicate
The 2010 series Scotch-Brite™ high performance microfibre wipes offer highly efficient, lint-free cleaning on virtually any surface, even delicate ones.
  • Highly efficient, lint-free cleaning on virtually any surface
  • Can be used with or without water, and with or without detergent
  • Extremely durable, wipes can resist up to 500 wash cycles
  • Machine washable up to 95 °C
Easily remove dust, dirt, oil, and grease with Scotch-Brite™ High Performance Microfibre Wipes 2010 Series. These high performance microfibre wipes offer highly efficient lint-free cleaningMore Product Information