Stampa…
Reusable earplugs, E-A-R™ Express™
Fornitore: 3M
EX01002 EX01001
111-1865EA 171 EUR
111-1865 MMMAEX01001
Reusable earplugs, E-A-R™ Express™
Tappi auricolari Inserti auricolari riutilizzabili
E-A-R™ Express™ Reusable Earplugs are recognisable yellow and blue earplugs with a foam tip that’s shaped and sized for a comfortable fit. They can reduce noise levels by 28 dB, and have a hygienic insertion grip.
  • Easy to slide gently into ears with slow recovery foam
  • Foam tip gives a comfortable and effective seal
  • Features a hygienic insertion grip
  • Recognisable blue stem and yellow tip for easy identification
Protect hearing at work or during leisure activities with the E-A-R™ Express™ Reusable Earplugs. Designed to slide gently into the ear, theMore Product Information