Stampa…
Safety Harnesses, XE200
Fornitore: 3M
7100316788 7100316790
MMMA7100316788EA 472 EUR
MMMA7100316788 MMMA7100316790
Safety Harnesses, XE200
Protezione anticaduta Imbragature di sicurezza
3M DBI-SALA ExoFit XE200 harness with rescue loops, automatic buckles, two sizes. X-shaped chest straps with central D-ring attachment point optimised position on the body. Revolver adjuster for the torso length for a quick, secure and comfortable fit of the harness.
  • Auto-locking quick connect
  • Length: 260 mm
  • Fall arrest loop placement: Back, front, shoulder
  • Product colour: grey
  • Integrated flat suspension trauma step loops
  • Reusable and repositionable lanyard holders for quick and easy parking of carabiners
The XE200 comfort harness consists of aMore Product Information