Stampa…
Microfil® Silicon Tubing
Fornitore: Merck
MIACTUB01
MILXMIACTUB01EA 0 EUR
MILXMIACTUB01
Microfil® Silicon Tubing
Tubazioni Tubazione di trasferimento
The silicon tubing is an additional accessory of the Microfil® filtration system. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. The push-fit design of the Microfil® funnels seals tightly to manifold supports without the need for clamps or O-rings. The tube is used to connect the pump to the manifold or filtration apparatus using the Tee connectors. The tubing is autoclavable, 5 m long, with an internal diameter of 9,5 mm.
The silicon tubing is an additional accessory of the Microfil® filtration system used in