Stampa…
Milli-Q® HX 7040/7080/7120/7150 SD Water Purification Systems
Fornitore: Merck
PR0GTXLCS1 ZLXLSD52040 ZLXLSD52080 ZLXLSD52150 ZLXLSD52120 PR0GTXLCS2
MILLPR0GTXLCS1EA 779 EUR
MILLPR0GTXLCS1 MILLZLXLSD52040 MILLZLXLSD52080 MILLZLXLSD52150 MILLZLXLSD52120 MILLPR0GTXLCS2
Milli-Q® HX 7040/7080/7120/7150 SD Water Purification Systems
Purificatori d'acqua Sistemi per acqua pura e ultrapura
Purifies up to 3000 L/day with integrated storage, distribution and remote monitoring.
  • Optimal combination of purification technologies for a reliable flow of high quality pure water
  • Integrated 140 L reservoir and pump for protected storage and distribution up to 20 L/min
  • State-of-the-art sustainable technologies for low and predictable running costs
  • Superior communications interface and modern data management capabilities
  • Customizable range of options and accessories to meet your specific requirements
  • MyMilli-Q™ Remote care online monitoring and service