Stampa…
Ultrafree®-MC Centrifugal Filter Units, with Microporous Hydrophilic PVDF (Durapore®) Membrane
Fornitore: Merck
UFC30DV00 UFC30HVNB UFC30VV00 UFC30GVNB UFC30HV00 UFC30GV00 UFC30VV25 UFC30GV25 UFC30DV25 UFC30HV25 UFC30GV0S UFC30DV0S UFC30SV00 UFC30HVIS
MILFUFC30DV00EA 525 EUR
MILFUFC30DV00 MILFUFC30HVNB MILFUFC30VV00 MILFUFC30GVNB MILFUFC30HV00 MILFUFC30GV00 516-0540 516-0538 MILFUFC30DV25 516-0539 MILFUFC30GV0S MILFUFC30DV0S MILFUFC30SV00 MILFUFC30HVIS
Ultrafree®-MC Centrifugal Filter Units, with Microporous Hydrophilic PVDF (Durapore®) Membrane
Filtri Filtri centrifughi
Ultrafree®-MC centrifugal devices are single-use, disposable PP filters used for removing particulates from aqueous biological solutions. Devices are available with a range of microporous membranes for fast filtration and highly reproducible performance.
  • Fast filtration and highly reproducible performance
Applications include clarification of aqueous and some solvent based samples, protein and nucleic acid sample preparation.