Stampa…
Millex® Hydromillex Syringe Filter
Fornitore: Merck
SLST025DM
MILFSLST025DMEA 977 EUR
MILFSLST025DM
Millex® Hydromillex Syringe Filter
Filtri Filtri per siringhe
The Hydromillex syringe filter is used downstream in a Technecium Generator to prevent particles of radioactive technecium from getting through to the collection vial where the liquid is collected.
  • Hydrophobic PTFE membrane has stripes of hydrophilic coating to allow simultaneous liquid and gas filtration
  • FDA registered medical device
  • RUO outside of US