Stampa…
Millex® Automation-Compatible Filter Units, with Nylon Membrane
Fornitore: Merck
SLHNBZ5NK SLHNBZ5NZ SLHNDZ5NK SLHNDZ5NZ SLGNDZ5NK SLGNDZ5NZ
MILFSLHNBZ5NKEA 3250 EUR
MILFSLHNBZ5NK MILFSLHNBZ5NZ MILFSLHNDZ5NK MILFSLHNDZ5NZ MILFSLGNDZ5NK MILFSLGNDZ5NZ
Millex® Automation-Compatible Filter Units, with Nylon Membrane
Filtri Filtri per siringhe
Automation compatible filter units are specially designed to provide efficient filtration for dissolution and other automated sample preparation applications.
  • Domed housing ensures reliable delivery of filters
  • Pressure-resistant housing resists bursting
  • Luer-Lok® connection optimised for precise alignment and fit
Automation compatible filter units are available in a variety of membrane types and pore sizes, including units with pre-filters for particulate-laden samples. Used in the process of drug dissolution testing, automated sample preparation,More Product Information