Stampa…
Millex® Syringe Filters, Polyethersulfone
Fornitore: Merck
SLGPX13NK SLGPX13NL SLGPR33RB SLHPR33RB SLHPR33RS
MILFSLGPX13NKEA 2020 EUR
MILFSLGPX13NK MILFSLGPX13NL MILFSLGPR33RB MILFSLHPR33RB MILFSLHPR33RS
Millex® Syringe Filters, Polyethersulfone
Filtri Filtri per siringhe
Syringe filters are single-use, membrane-based devices used for the removal of particulate impurities from liquid and gas samples prior to analysis by methods such as HPLC, ion chromatography, gas chromatography, ICP, and dissolution testing. Proper filtration of samples improves the quality and consistency of analytical results and decreases instrument downtime.
  • Non-sterile Millex® syringe filters contain Millipore Express™ PLUS polyethersulfone (PES) membrane
  • Enables fast filtration of aqueous and mild organic solutions, 0,22 μm pore size diameters to