Stampa…
Stericup®, Steritop™ Bottle Top Vacuum Filtration Systems, PES Membrane
Fornitore: Merck
SCGPU05RE SCGPU02RE SCGPS01RE SCGPT01RE SCGPS02RE SCGPT02RE SCGPS05RE S2GPT10RE S2GPT02RE S2GPT01RE S2GPT05RE S2VPU02RE S2VPU11RE
515-0031EA 227 EUR
515-0031 515-0030 515-0021 515-0025 515-0022 515-0026 515-0023 MILFS2GPT10RE MILFS2GPT02RE MILFS2GPT01RE MILFS2GPT05RE MILFS2VPU02RE MILFS2VPU11RE
Stericup®, Steritop™ Bottle Top Vacuum Filtration Systems, PES Membrane
Filtri Filtri per bottiglie
Ideal for sterilising tissue culture media, buffers, or other aqueous solutions. The no tip, tapered receiver flask and compact profile of the Stericup® design improve stability and make gripping the receiver easier.
  • Connections: Hose barb vacuum inlet
  • Max. operating temperature: 45 °C
  • Sterile
The bottom of the flask is slightly recessed so that capped flasks can be stacked for storage. A small tab inside the funnel secures a prefilter in place during filtration. For added protection against contamination, the threads of the bottle cap are slightlyMore Product Information