Stampa…
Stericup®, Steritop™ Bottle Top Vacuum Filtration Systems, PVDF Membrane
Fornitore: Merck
SCGPU11RE SC00B02RE SC00B10RE SCGVU02RE SCGVT05RE SCHVU01RE SCHVU11RE SCGVU11RE S2GPU10RE S2GPU11RE S2GPU02RE S2HVU11RE S2GPU01RE S2HVU05RE S200B05RE S2GVU10RE S200B02RE S200B10RE S2GPU05RE S2HVU01RE S2HVU02RE
515-0033EA 0 EUR
515-0033 MILFSC00B02RE MILFSC00B10RE 515-0037 MILFSCGVT05RE 515-0041 515-0045 515-0040 MILFS2GPU10RE MILFS2GPU11RE MILFS2GPU02RE MILFS2HVU11RE MILFS2GPU01RE MILFS2HVU05RE MILFS200B05RE MILFS2GVU10RE MILFS200B02RE MILFS200B10RE MILFS2GPU05RE MILFS2HVU01RE MILFS2HVU02RE
Stericup®, Steritop™ Bottle Top Vacuum Filtration Systems, PVDF Membrane
Filtri Filtri per bottiglie
Ideal for sterilising tissue culture media, buffers, or other aqueous solutions. The no tip, tapered receiver flask and compact profile of the Stericup® design improve stability and make gripping the receiver easier.
  • Connections: Hose barb vacuum inlet
  • Max. operating temperature: 45 °C
  • Sterile
The bottom of the flask is slightly recessed so that capped flasks can be stacked for storage. A small tab inside the funnel secures a prefilter in place during filtration. For added protection against contamination, the threads of the bottle cap are slightlyMore Product Information