Stampa…
MultiScreenHTS Filter Plates with Hydrophilic MCE Membrane, 96-Well
Fornitore: Merck
MAHAN4550 MAHAS4510 MSHAN4510 MSHAS4510 MSHAN4550 MSHAN4B50 MSHAS4B10
736-0183EA 1450 EUR
736-0183 736-0184 MILFMSHAN4510 MILFMSHAS4510 MILFMSHAN4550 MILFMSHAN4B50 MILFMSHAS4B10
MultiScreenHTS Filter Plates with Hydrophilic MCE Membrane, 96-Well
Micropiastre Piastre filtranti
MultiScreenHTS plates are specifically developed for high-throughput use with automated work stations. Rigid sidewalls improve handling and ample surfaces are provided for bar code labels.
  • Hydrophilic mixed cellulose esters (MCE) membrane
Wells are individually sealed to prevent incubation crosstalk and the underdrain is removable for access to the filters.
MultiScreen HTS 96-well filtration plates are used to filter samples and perform entire procedures, from cell growth to scintillation counting within the same plate. The plates come in manyMore Product Information