Stampa…
Membrane filters, Fluoropore™
Fornitore: Merck
FHUP04700 FALP01300 FALP02500 FALP03700 FALP04700 FALP09050 FALP14250 FGLP01300 FGLP02500 FGLP09050 FGLP14250 FHLP01300 FHLP02500 FHLP03700 FHLP09050 FHLP14250 FSLW02500 FSLW04700 FSLW09025 FSLW14200 FGLP04700 FHLP04700 FSLW03700 FSLW00010 FGLP00010 FMLW04700 FSLW05800 FTHP04750 FSLW06050 FGLP03700
MILFFHUP04700EA 733 EUR
MILFFHUP04700 MILFFALP01300 MILFFALP02500 MILFFALP03700 MILFFALP04700 MILFFALP09050 MILFFALP14250 MILFFGLP01300 MILFFGLP02500 MILFFGLP09050 MILFFGLP14250 MILFFHLP01300 MILFFHLP02500 MILFFHLP03700 MILFFHLP09050 MILFFHLP14250 MILFFSLW02500 MILFFSLW04700 MILFFSLW09025 MILFFSLW14200 512-0246 512-0248 MILFFSLW03700 MILFFSLW00010 MILFFGLP00010 MILFFMLW04700 MILFFSLW05800 MILFFTHP04750 MILFFSLW06050 MILFFGLP03700
Membrane filters, Fluoropore™
Filtri Membrane filtranti
These white Fluoropore™ membrane are hydrophobic, PTFE polymer membrane bonded to a high density PE support and provide broad chemical compatibility, high flow rates and consistency.
  • PTFE with and without backing material
  • Biologically and chemically inert
  • High porosity yields high flow rates
  • Can be sterilised by ethylene oxide or autoclave
  • Available for filtration of aqueous and organic based samples
They are used for clarifying acids, bases and solvents, air monitoring, filtering and venting gases and uv spectroscopy.
Filter surface: plain