Stampa…

Visualizer and Visualizer EC Western Blot Detection Kits

Fornitore: Merck

64-201 64-202BP 64-202 64-201BP
MILF64-201EA 533 EUR
MILF64-201 MILF64-202BP MILF64-202 MILF64-201BP
Visualizer and Visualizer EC Western Blot Detection Kits
Reagenti per elettroforesi Reagenti per Western Blotting

The Visualizer Western Blot Detection kit is designed for the sensitive detection of proteins in chemiluminescent Western blots.


  • Storage: Stable for 18 months at 4 °C

  • Application: For use on immunoblots


The reagents in this kit have been matched to optimise the range and sensitivity of detection using various antibodies in Western blot assays.

Components: Goat anti-mouse IgG, goat anti-rabbit IgG, HRP conjugate Visualizer detection reagent A Visualizer detection reagent B.



Visualizer EC western blot detection kits are designed with a high glow reaction chemiluminescent substrate with outstanding sensitivity, long-lasting signal and low cost. Visualizer EC offers the highest signal-to-noise ratios of any existing luminol peroxidase formulations, resulting in high sensitivity and low backgrounds.


Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR