Stampa…
Laboratory Glassware Washers, Large Capacity, PLW 8616
Fornitore: MIELE
10880700 10928990 10931330 10941870 10880860 10941940 11176500 11176510 11176530 11176540 10880710 11332020
MIEL10880700EA 0 EUR
MIEL10880700 MIEL10928990 MIEL10931330 MIEL10941870 MIEL10880860 MIEL10941940 MIEL11176500 MIEL11176510 MIEL11176530 MIEL11176540 MIEL10880710 MIEL11332020
Laboratory Glassware Washers, Large Capacity, PLW 8616
Lavavetreria
The PLW 8616 laboratory washer can accommodate 216 laboratory flasks, 588 vials, or 294 pipettes. The large glassware washer is flexible and simple to use, it has modular load carriers and SimpleLoad system. This efficient unit has a double door, 900 mm wide, a usable capacity of 351 litres. PLW 8616 provides reliable results, high pump performance with a variable-speed pump and features spray arm monitoring and a conductivity meter.
  • Innovative 2-pump system for maximum performance
  • New flexible and modular (SimpleLoad) racking system
  • Excellent drying