Stampa…
Masterflex® Transfer Tubing Spool, PharMed® BPT, Avantor®
Fornitore: Avantor Fluid Handling
MFLX96900-06EA 2710 EUR
MFLX96900-06
Masterflex® Transfer Tubing Spool, PharMed® BPT, Avantor®
Tubazioni Tubazione di trasferimento
Protect your light-sensitive samples.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. TemperatureMore Product Information