Stampa…
Masterflex® Ismatec® Microbore Pump Tubing, Puri-Flex®, Avantor®
Fornitore: Avantor Fluid Handling
MFLX96418-10EA 86.7 EUR
MFLX96418-10 MFLX96418-12 MFLX96418-14 MFLX96418-16 MFLX96418-18 MFLX96418-22 MFLX96418-24 MFLX96418-26 MFLX96418-28 MFLX96418-30 MFLX96418-34 MFLX96418-36 MFLX96418-40 MFLX96418-42 MFLX96418-44 MFLX96418-46 MFLX96418-48 MFLX96418-32
Masterflex® Ismatec® Microbore Pump Tubing, Puri-Flex®, Avantor®
Tubazioni Tubo per pompa peristaltica
Long life tubing for pharma and bioprocess applications.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
Applications: General-purpose lab use requiring high performance and long life
Characteristics: Exceptional tensile and tear strength, goodMore Product Information