Stampa…
Masterflex® Transfer Tubing, Peroxide-Cured PTFE, Red, Avantor®
Fornitore: Avantor Fluid Handling
96130-02 96130-04
MFLX96130-02EA 68 EUR
MFLX96130-02 MFLX96130-04
Masterflex® Transfer Tubing, Peroxide-Cured PTFE, Red, Avantor®
Tubazioni Tubazione di trasferimento
Quickly identify lines.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Ideal for dedicated or multiple lines—provides quick and easy line identification.
Characteristics: Chemically inert, nontoxic, and virtually nonporous. Opaque.
Certifications: None.
Temperature range: -400 to 500 °F (-240 to 260More Product Information