Stampa…
Masterflex® Transfer Tubing Spool, Polytetrafluoroethylene (PTFE), Avantor®
Fornitore: Avantor Fluid Handling
95231-02 95231-06
MFLX95231-00EA 978 EUR
MFLX95231-00 MFLX95231-08 MFLX95231-10 MFLX95231-12 MFLX95231-14 MFLX95231-02 MFLX95231-06
Masterflex® Transfer Tubing Spool, Polytetrafluoroethylene (PTFE), Avantor®
Tubazioni Tubazione di trasferimento
Available in spooled rolls for increased versatility.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP.More Product Information