Stampa…
Masterflex® Transfer Tubing, Kynar Flex™ 2750 PVDF, Avantor®
Fornitore: Avantor Fluid Handling
MFLX95100-01EA 93.9 EUR
MFLX95100-01 MFLX95100-03 MFLX95100-05 MFLX95100-07 MFLX95100-09 MFLX95100-11 MFLX95100-13 MFLX95100-15 MFLX95100-19 MFLX95100-21
Masterflex® Transfer Tubing, Kynar Flex™ 2750 PVDF, Avantor®
Tubazioni Tubazione di trasferimento
Excellent chemical resistance and UV stable.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: High purity and temperature, UV sensitive, or aggressive chemical fluid transfer.
Characteristics: Excellent chemical resistance, low TOC, rigid tubing works well with compression and over-molded fittingsMore Product Information