Stampa…
Masterflex® L/S® Digital Modular Dispensing Drives, Avantor®
Fornitore: Avantor Fluid Handling
MFLX77301-40-EUEA 4640 EUR
MFLX77301-40-EU MFLX77301-50-EU MFLX77301-60-EU MFLX77301-70-EU MFLX77301-40-IT MFLX77301-50-IT MFLX77301-60-IT MFLX77301-70-IT
Masterflex® L/S® Digital Modular Dispensing Drives, Avantor®
Pompe Azionamenti di pompe Peristaltic Pump Drives
Precision digital drives offer three dispensing modes and precision low-maintenance motors— modular format lets you place motor and controller where convenient.
  • Flow rate 0.0012 to 3400 ml/min with L/S —flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for