Stampa…
Masterflex® Mechanically Coupled Self-Priming Centrifugal Pumps, Avantor®
Fornitore: Avantor Fluid Handling
75505-71 75505-72 75505-74
MFLX75505-71EA 1430 EUR
MFLX75505-71 MFLX75505-72 MFLX75505-74
Masterflex® Mechanically Coupled Self-Priming Centrifugal Pumps, Avantor®
Pompe Pompe Centrifughe
Chemical-resistance pump heads provide dependable performance and continuous service.
  • Mechanically close-coupled design saves space and simplifies installation and maintenance
  • Back pullout construction — assembly and overhaul of impeller and seal without distorting suction and discharge connections
  • Top centerline discharge and foot support under casing ensure self-venting and reduce misalignment from pipe loads
The mechanically coupled centrifugal pumps, constructed of 304 stainless steel, are ideal for food and pharmaceutical processing plants, asMore Product Information