Stampa…
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, EPDM, Avantor®
Fornitore: Avantor Fluid Handling
30537-04 30537-08 30537-20 30537-22 30537-24 30537-28 30537-40 30537-42 30537-44 30537-48 30537-60 30537-68
MFLX30537-06EA 68.7 EUR
MFLX30537-06 MFLX30537-26 MFLX30537-46 MFLX30537-62 MFLX30537-64 MFLX30537-66 MFLX30537-80 MFLX30537-82 MFLX30537-84 MFLX30537-86 MFLX30537-88 MFLX30537-04 MFLX30537-08 MFLX30537-20 MFLX30537-22 MFLX30537-24 MFLX30537-28 MFLX30537-40 MFLX30537-42 MFLX30537-44 MFLX30537-48 MFLX30537-60 MFLX30537-68
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, EPDM, Avantor®
Guarnizioni Guarnizioni
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derivedMore Product Information