Stampa…
Masterflex® Strainer Gasket for Low-Cost In-Line Strainer Systems, Avantor®
Fornitore: Avantor Fluid Handling
29595-81 29595-83 29595-85 29595-87 29595-89 29595-91
MFLX29595-81EA 7.4 EUR
MFLX29595-81 MFLX29595-83 MFLX29595-85 MFLX29595-87 MFLX29595-89 MFLX29595-91
Masterflex® Strainer Gasket for Low-Cost In-Line Strainer Systems, Avantor®
Filtri Strainers
Easily customize strainer system to suit your application.
  • Choose from either EDPM or Viton® gaskets based on chemical compatibility requirements
Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate, and large. Modify or change components of the same class to meet the demands of your application.
Low-Profile Strainers are ideal for small-scale low-volume filtration. Use in research applications, cleaning equipment, and laboratory equipment.
Intermediate StrainersMore Product Information