Stampa…
Parker Hannifin Domnick Hunter Air Purification Systems
Fornitore: Avantor Fluid Handling
29587-03 29587-07 29587-11 29587-13 29587-15 29587-23
MFLX29587-03EA 149 EUR
MFLX29587-03 MFLX29587-07 MFLX29587-11 MFLX29587-13 MFLX29587-15 MFLX29587-23
Parker Hannifin Domnick Hunter Air Purification Systems
Purificatori d'aria
Advanced airflow technology makes this the most energy efficient compressed air filtration system available.
  • Prefilters and microfilters feature an internal float drain that eases installation and saves initial cost
  • Incident monitor (included on most models) alerts you to high differential pressure and need for filter element change
  • Housing is fully corrosion protected and carries a 10-year warranty
These modular filters are the high-efficiency answer to your compressed air filtration needs. The filter housing is Alocrom-treated aluminum with a high-More Product Information