Stampa…
Masterflex® L/S® Precision Variable-Speed Console Drives, Avantor®
Fornitore: Avantor Fluid Handling
MFLX07528-10-EUEA 2180 EUR
MFLX07528-10-EU MFLX07528-20-EU MFLX07528-30-EU MFLX07528-10-IT MFLX07528-20-IT MFLX07528-30-IT
Masterflex® L/S® Precision Variable-Speed Console Drives, Avantor®
Pompe Azionamenti di pompe Peristaltic Pump Drives
Don’t lose control of your metering applications—digital display gives you repeatable control.
  • Flow rate 0.06 to 3400 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Three digit LED display shows motor rpm — confirm speed setting at a glance
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for multichannel applications
  • Separate speed and power (on/off) control — maintain speed setting