Stampa…
American Machine & Tool Motor Foot Bracket
Fornitore: Avantor Fluid Handling
07127-80
MFLX07127-80EA 34.1 EUR
MFLX07127-80
American Machine & Tool Motor Foot Bracket
Pompe Motori
Create your own motor mounting with the hardware below.
  • Easily mount your American Machine & Tool Motor NEMA Type 56 C-face motors
Motor Foot Bracket provides a base for NEMA Type 56 C-face motors.