Stampa…
Masterflex® L/S® Standard Pump Heads for High-Performance Precision Tubing, Avantor®
Fornitore: Avantor Fluid Handling
MFLX07015-20EA 298 EUR
MFLX07015-20 MFLX07015-21 MFLX07015-52 MFLX07024-20 MFLX07024-21 MFLX07024-52 MFLX07035-20 MFLX07035-21 MFLX07036-30 MFLX07036-31
Masterflex® L/S® Standard Pump Heads for High-Performance Precision Tubing, Avantor®
Pompe Teste della pompa Peristaltic Pump Heads
Ideal for fluid transfer applications requiring high accuracy and repeatability.
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design with same side tubing entry/exit—perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Stack up to four heads depending on drive recommendations
This Standard pump head accepts high-performance precision tubing— choose these pump heads for better pressure generation, suction lift, viscous fluidMore Product Information