Stampa…
Chemfluor® ETFE Transfer Pump Tubing
Fornitore: Avantor Fluid Handling
06488-15
MFLX06488-07EA 299 EUR
MFLX06488-07 MFLX06488-08 MFLX06488-09 MFLX06488-10 MFLX06488-11 MFLX06488-12 MFLX06488-14 MFLX06488-15
Chemfluor® ETFE Transfer Pump Tubing
Tubazioni Tubazione di trasferimento
Meets the demands of various industries.
  • Ideal for general-purpose, single-use, and extended fluid path applications
Tubing is translucent and slightly flexible. Tubing is versatile and can be used in several applications, including lab, chemical, electrical, mechanical, and automotive applications. Ideal for biochromatography-will not denature sensitive biomolecules. ETFE exhibits excellent mechanical resistance and is more chemically inert than SS tubing. Withstands pressures up to 3000 psi and can be used continuously at temperatures up to 300 °F (1More Product Information