Stampa…
Masterflex® I/P® Precision Pump Tubing, Chem-Durance® Bio, Avantor®
Fornitore: Avantor Fluid Handling
MFLX06442-26EA 706 EUR
MFLX06442-26 MFLX06442-73 MFLX06442-82
Masterflex® I/P® Precision Pump Tubing, Chem-Durance® Bio, Avantor®
Tubazioni Tubo per pompa peristaltica
Enhanced pumping life and durability—especially under pressure.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
Masterflex® Chem-Durance® Bio Precision pump tubingMore Product Information