Stampa…
Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Fornitore: Avantor Fluid Handling
MFLX06434-35EA 199 EUR
MFLX06434-35 MFLX06434-36 MFLX06434-24
Masterflex® L/S® High-Performance Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Tubazioni Tubo per pompa peristaltica
Longest life of any C-Flex® formulation.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-