Stampa…
Masterflex® Transfer Tubing, Tygon® Tygon® S3™ E-3603, Avantor®
Fornitore: Avantor Fluid Handling
06407-99
MFLX06407-70EA 39.1 EUR
MFLX06407-70 MFLX06407-71 MFLX06407-72 MFLX06407-73 MFLX06407-74 MFLX06407-75 MFLX06407-76 MFLX06407-77 MFLX06407-78 MFLX06407-79 MFLX06407-80 MFLX06407-81 MFLX06407-82 MFLX06407-83 MFLX06407-84 MFLX06407-85 MFLX06407-86 MFLX06407-87 MFLX06407-88 MFLX06407-89 MFLX06407-90 MFLX06407-91 MFLX06407-92 MFLX06407-93 MFLX06407-94 MFLX06407-95 MFLX06407-96 MFLX06407-98 MFLX06407-99
Masterflex® Transfer Tubing, Tygon® Tygon® S3™ E-3603, Avantor®
Tubazioni Tubazione di trasferimento
Pliable formulation for sharp radius bending.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. It offers long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, and is nontoxic.
FDA compliant and made according to GMP.