Stampa…
CPC® Plastic Quick-Disconnect Fittings, Complete Couplings
Fornitore: Avantor Fluid Handling
MFLX06360-90EA 29.8 EUR
MFLX06360-90 MFLX06360-91 MFLX31305-00 MFLX31305-05 MFLX31306-05 MFLX31305-05BLK MFLX31306-05BLK MFLX06360-90BLK MFLX06360-91BLK MFLX31305-00BLK
CPC® Plastic Quick-Disconnect Fittings, Complete Couplings
Raccordi e connettori
Fittings connect in a snap.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Acetal fittings are ideal for use with corrosive fluids
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
CompleteMore Product Information