Stampa…
Super Adjustable™ Super Erecta® stem castor carts
Fornitore: Metro International
5A356EC 5A336BC 5A356BC A366BC
METO5A356ECEA 1140 EUR
METO5A356EC METO5A336BC METO5A356BC METOA366BC
Super Adjustable™ Super Erecta® stem castor carts
Arredi Ripiani
Designed to work in conjunction with the entire Super Erecta® system of shelves and accessories. These Super adjustable wire shelves have a unique corner release system which allows them to be adjusted quickly and easily without the use of tools.
  • Allow for quick and easy adjustability
  • Strong and versatile
  • 5- or 4-tier models
  • Resilient rubber or polyurethane castors with donut bumpers
Shelf has a sleek chrome finish that makes it perfect for dry storage areas, while the open-wire construction minimises dust accumulation and allows a free circulation ofMore Product Information