Stampa…
Collection containers, Corning®
Fornitore: Corning
91-001-MB 91-100-30 91-200-01 91-200-02 91-200-05 91-200-10 91-200-20 91-200-36 91-200-39 91-200-41 91-200-42 91-200-43 91-200-45 91-200-47 91-200-48 91-200-82 91-200-83
MDTC91-001-MBEA 0 EUR
MDTC91-001-MB MDTC91-100-30 MDTC91-200-01 MDTC91-200-02 MDTC91-200-05 MDTC91-200-10 MDTC91-200-20 MDTC91-200-36 MDTC91-200-39 MDTC91-200-41 MDTC91-200-42 MDTC91-200-43 MDTC91-200-45 MDTC91-200-47 MDTC91-200-48 MDTC91-200-82 MDTC91-200-83
Collection containers, Corning®
Sacchetti Sacchetti di terreni
Corning® collection containers are fabricated to match specific application requirements, and are available in a variety of sizes and connector configurations.
  • Single-use bags
  • Material ULDPE
  • Designed to be practical and cost-effective alternatives to rigid-walled containers
  • Ideal for biopharmaceutical applications
Unless otherwise noted in container port descriptions, all port tubing components are chemically resistant, heat sealable flexible tubing.
Port 1: 60 L × ¹/₂ Int.Ø ×³/₄ Ext.Ø tubing, Male MPX connector, Female MPC cap, pinch clamp.