Stampa…

Cloramfenicolo, in polvere

Fornitore: Corning

61-239-RI
MDTC61-239-RIEA 105.14 EUR
MDTC61-239-RI
Cloramfenicolo, in polvere
Cloramfenicolo

Chloramphenicol is a common antibiotic which inhibits microbial protein synthesis by preventing peptide bond formation. It is effective against Gram-positive and Gram-negative bacteria.


Confezione: Shipping temperature: Ambient

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00078159
CAS Number: 56-75-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR