Stampa…

Sodio cloruro solution_SC 5M DNAse_RNAse_PROTEase_free, Corning®

Fornitore: Corning

Synonyms: Saline, NaCl, Sea salt, Halite

46-032-CV
MDTC46-032-CVEA 299.05 EUR
MDTC46-032-CV
Sodio cloruro solution_SC 5M DNAse_RNAse_PROTEase_free, Corning®
Sodio cloruro

The 5 M NaCl is a common molecular biology reagent.


All production lots are tested to ensure that they are free of RNAse. This product is used to control osmolarity and conductivity, among other factors.

Formula: NaCl
Peso molecolare: 58.44 g/mol
Punto di ebollizione: 1413
Punto di fusione: 801
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR