Stampa…

Acido cloridrico 0.714 mol/l (N/1.4)

Fornitore: PROLABO CMP
Attenzione

Synonyms: Hydrochloric acid solution, Muriatic acid

MC307904.6010
MC307904.6010EA 0 EUR
MC307904.6010
Acido cloridrico 0.714 mol/l (N/1.4)
Acido cloridrico
Formula: HCl
Peso molecolare: 36.46 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR